Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ugt-18
Entrez Gene ID
179759
Gene Name
UDP-glucuronosyltransferase
Gene Aliases
CELE_ZC443.5
Chromosome Location
Chr.V: 12820260 - 12822778 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073808.6 NP_506209.2 6 - 7 1161 94

View Full Product Details