Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ugt-15
Entrez Gene ID
183462
Gene Name
UDP-glucuronosyltransferase
Gene Aliases
CELE_C44H9.1
Chromosome Location
Chr.V: 12827519 - 12830114 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073810.6 NP_506211.1 1 - 2 462 117

View Full Product Details