Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nid-1
Entrez Gene ID
179770
Gene Name
protein-coding
Gene Aliases
CELE_F54F3.1
Chromosome Location
Chr.V: 12910107 - 12917221 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001269536.4 NP_001256465.1 9 - 10 2689 127
NM_001269537.3 NP_001256466.1 9 - 10 2695 127
NM_001269538.3 NP_001256467.1 9 - 10 2695 127

View Full Product Details