Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srh-18
Entrez Gene ID
183633
Gene Name
Serpentine Receptor, class H
Gene Aliases
CELE_C50B6.5
Chromosome Location
Chr.V: 13318452 - 13320225 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073900.2 NP_506301.1 2 - 3 315 148

View Full Product Details