Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srv-10
Entrez Gene ID
186173
Gene Name
Serpentine Receptor, class V
Gene Aliases
CELE_F53F1.9
Chromosome Location
Chr.V: 13423908 - 13425266 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073928.1 NP_506329.1 3 - 4 384 97

View Full Product Details