Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
swan-1
Entrez Gene ID
179876
Gene Name
WD_REPEATS_REGION domain-containing protein
Gene Aliases
CELE_F53C11.8
Chromosome Location
Chr.V: 13804391 - 13807237 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_074017.7 NP_506418.1 5 - 6 1107 70

View Full Product Details