Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nrf-5
Entrez Gene ID
179879
Gene Name
Nose resistant to fluoxetine protein 5
Gene Aliases
CELE_F55B12.5
Chromosome Location
Chr.V: 13826959 - 13831074 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_074023.8 NP_506424.2 3 - 4 631 96

View Full Product Details