Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- D1086.3
- Entrez Gene ID
-
179907
- Gene Name
- DUF19 domain-containing protein
- Gene Aliases
-
CELE_D1086.3
- Chromosome Location
-
Chr.V: 14090462 - 14093986 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001383449.1 |
|
5 - 6 |
1014 |
64 |
- Gene Symbol
- him-5
- Entrez Gene ID
-
183932
- Gene Name
- protein-coding
- Gene Aliases
-
CELE_D1086.4
- Chromosome Location
-
Chr.V: 14090426 - 14092454 on Build WBcel235
View Full Product Details