Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sre-8
Entrez Gene ID
185561
Gene Name
Serpentine receptor class epsilon-8
Gene Aliases
CELE_F40G12.1
Chromosome Location
Chr.V: 14257113 - 14259231 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_074127.3 NP_506528.1 5 - 6 889 73

View Full Product Details