Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srm-4
Entrez Gene ID
185899
Gene Name
Serpentine Receptor, class M
Gene Aliases
CELE_F47B8.7
Chromosome Location
Chr.V: 14333416 - 14334806 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_074153.4 NP_506554.1 4 - 5 818 98

View Full Product Details