Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
set-31
Entrez Gene ID
179939
Gene Name
SET (trithorax/polycomb) domain containing
Gene Aliases
CELE_C15H11.5
Chromosome Location
Chr.V: 14417903 - 14420144 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_074168.7 NP_506569.1 7 - 8 1280 82

View Full Product Details