Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gcy-14
Entrez Gene ID
191647
Gene Name
Receptor-type guanylate cyclase gcy-14
Gene Aliases
CELE_ZC412.2
Chromosome Location
Chr.V: 14863134 - 14871530 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_074259.4 NP_506660.2 8 - 9 1934 70

View Full Product Details