Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nmr-2
Entrez Gene ID
180002
Gene Name
protein-coding
Gene Aliases
CELE_T01C3.10
Chromosome Location
Chr.V: 15006341 - 15011660 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001269711.3 NP_001256640.1 9 - 10 1545 127
GenBank mRNA AF318614.2 - 9 - 10 1521 127

View Full Product Details