Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cyp-34A2
Entrez Gene ID
188400
Gene Name
CYtochrome P450 family
Gene Aliases
CELE_T10H4.11
Chromosome Location
Chr.V: 15288777 - 15291107 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_074387.2 NP_506788.1 6 - 7 1392 85

View Full Product Details