Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srx-48
Entrez Gene ID
188961
Gene Name
G_PROTEIN_RECEP_F1_2 domain-containing protein
Gene Aliases
CELE_T26H8.2
Chromosome Location
Chr.V: 15302371 - 15303725 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_074392.2 NP_506793.2 5 - 6 800 136

View Full Product Details