Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srx-58
Entrez Gene ID
183007
Gene Name
G_PROTEIN_RECEP_F1_2 domain-containing protein
Gene Aliases
CELE_C29F3.6
Chromosome Location
Chr.V: 15352676 - 15353985 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_074408.4 NP_506809.3 2 - 3 331 109

View Full Product Details