Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
fbxa-95
Entrez Gene ID
185071
Gene Name
F-box domain-containing protein
Gene Aliases
CELE_F28F8.4
Chromosome Location
Chr.V: 15571474 - 15572715 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_074470.4 NP_506871.3 2 - 3 198 76

View Full Product Details