Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
haf-3
Entrez Gene ID
180056
Gene Name
HAlF transporter (PGP related)
Gene Aliases
CELE_F57A10.3
Chromosome Location
Chr.V: 15760413 - 15766172 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_074526.7 NP_506927.2 4 - 5 501 72

View Full Product Details