Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cdr-1
Entrez Gene ID
180064
Gene Name
CaDmium Responsive
Gene Aliases
CELE_F35E8.11
Chromosome Location
Chr.V: 15921309 - 15922820 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_074585.9 NP_506986.1 5 - 6 749 84
GenBank mRNA AF301606.1 - 5 - 6 697 84

View Full Product Details