Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srw-53
Entrez Gene ID
188147
Gene Name
G_PROTEIN_RECEP_F1_2 domain-containing protein
Gene Aliases
CELE_T05G11.7
Chromosome Location
Chr.V: 15943425 - 15944854 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_074593.1 NP_506994.1 3 - 4 884 88

View Full Product Details