Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srx-43
Entrez Gene ID
188374
Gene Name
Serpentine receptor class X-43
Gene Aliases
CELE_T10C6.3
Chromosome Location
Chr.V: 16017971 - 16020066 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_074621.3 NP_507022.2 2 - 3 400 101

View Full Product Details