Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
oac-6
Entrez Gene ID
180094
Gene Name
NRF domain-containing protein
Gene Aliases
CELE_C31A11.5
Chromosome Location
Chr.V: 16302717 - 16306159 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_074717.5 NP_507118.1 9 - 10 1940 117

View Full Product Details