Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
str-15
Entrez Gene ID
188292
Gene Name
Seven TM Receptor
Gene Aliases
CELE_T08G3.1
Chromosome Location
Chr.V: 16457269 - 16458814 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_074761.2 NP_507162.2 3 - 4 775 78

View Full Product Details