Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
str-61
Entrez Gene ID
184475
Gene Name
Seven TM Receptor
Gene Aliases
CELE_F14F8.1
Chromosome Location
Chr.V: 16669339 - 16671290 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001356771.1 NP_001343590.1 1 - 2 491 165

View Full Product Details