Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
clec-38
Entrez Gene ID
188900
Gene Name
C-type LECtin
Gene Aliases
CELE_T25E12.10
Chromosome Location
Chr.V: 16733271 - 16735618 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_074832.3 NP_507233.2 1 - 2 242 75

View Full Product Details