Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
clec-29
Entrez Gene ID
180117
Gene Name
C-type LECtin
Gene Aliases
CELE_T25E12.9
Chromosome Location
Chr.V: 16737765 - 16739200 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_074834.6 NP_507235.2 1 - 2 193 144

View Full Product Details