Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
dhs-24
Entrez Gene ID
180306
Gene Name
DeHydrogenases, Short chain
Gene Aliases
CELE_Y60A3A.10
Chromosome Location
Chr.V: 19934767 - 19946622 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_075459.8 NP_507860.3 1 - 2 100 62

View Full Product Details