Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
lgc-55
Entrez Gene ID
180312
Gene Name
Ligand-Gated ion Channel
Gene Aliases
CELE_Y113G7A.5
Chromosome Location
Chr.V: 20023046 - 20036667 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_075469.6 NP_507870.2 7 - 8 1091 73

View Full Product Details