Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srxa-6
Entrez Gene ID
189273
Gene Name
Serpentine Receptor, class XA
Gene Aliases
CELE_W07A8.5
Chromosome Location
Chr.V: 20734958 - 20736180 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_075604.2 NP_508005.1 2 - 3 434 136

View Full Product Details