Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sor-3
Entrez Gene ID
180391
Gene Name
Sop-2-related protein 3
Gene Aliases
CELE_C04E7.2
Chromosome Location
Chr.X: 353619 - 357939 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_075689.6 NP_508090.2 6 - 7 695 97

View Full Product Details