Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mrp-1
Entrez Gene ID
180409
Gene Name
protein-coding
Gene Aliases
CELE_F57C12.5
Chromosome Location
Chr.X: 576312 - 587898 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029548.6 NP_001024719.2 2 - 3 304 97
NM_001038464.4 NP_001033553.1 2 - 3 310 97
NM_075721.9 NP_508122.2 3 - 4 569 97
NM_171938.8 NP_741702.1 2 - 3 300 97

View Full Product Details