Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- sms-2
- Entrez Gene ID
-
180445
- Gene Name
- Phosphatidylcholine:ceramide cholinephosphotransferase 2
- Gene Aliases
-
CELE_F53H8.4
- Chromosome Location
-
Chr.X: 944826 - 948885 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_075781.5 |
NP_508182.1 |
1 - 2 |
124 |
61 |
View Full Product Details