Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sms-2
Entrez Gene ID
180445
Gene Name
Phosphatidylcholine:ceramide cholinephosphotransferase 2
Gene Aliases
CELE_F53H8.4
Chromosome Location
Chr.X: 944826 - 948885 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_075781.5 NP_508182.1 4 - 5 700 89

View Full Product Details