Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
arg-1
Entrez Gene ID
180501
Gene Name
Delta-like protein
Gene Aliases
CELE_F31A9.3
Chromosome Location
Chr.X: 1786907 - 1788405 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029444.1 NP_001024615.1 5 - 6 923 54

View Full Product Details