Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pqn-15
Entrez Gene ID
180705
Gene Name
Prion-like-(Q/N-rich)-domain-bearing protein
Gene Aliases
CELE_C24A8.3
Chromosome Location
Chr.X: 4297869 - 4306961 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001380945.1 NP_001367558.1 5 - 6 779 74

View Full Product Details