Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C52B9.8
Entrez Gene ID
182830
Gene Name
Uncharacterized protein
Gene Aliases
CELE_C52B9.8
Chromosome Location
Chr.X: 4307770 - 4311768 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001355087.2 NP_001342016.1 1 - 2 131 80

View Full Product Details