Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ifa-4
Entrez Gene ID
180765
Gene Name
Intermediate filament protein ifa-4
Gene Aliases
CELE_K05B2.3
Chromosome Location
Chr.X: 4912308 - 4915991 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_076435.9 NP_508836.5 6 - 7 1054 84

View Full Product Details