Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lfi-1
Entrez Gene ID
180771
Gene Name
protein-coding
Gene Aliases
CELE_ZC8.4
Chromosome Location
Chr.X: 4976830 - 4993242 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001375027.2 NP_001362033.1 14 - 15 5533 95
NM_001375029.2 NP_001362032.1 13 - 14 5326 95
NM_001375030.2 NP_001362152.1 13 - 14 5484 95
NM_001375031.2 NP_001362153.1 12 - 13 5234 95
NM_001383570.1 14 - 15 5837 95
NM_076447.6 NP_508848.2 15 - 16 5871 95

View Full Product Details