Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lfi-1
Entrez Gene ID
180771
Gene Name
protein-coding
Gene Aliases
CELE_ZC8.4
Chromosome Location
Chr.X: 4976830 - 4993242 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001375031.2 NP_001362153.1 1 - 2 142 72
NM_076447.6 NP_508848.2 4 - 5 779 72

View Full Product Details