Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
acn-1
Entrez Gene ID
180780
Gene Name
Inactive angiotensin-converting enzyme-related protein
Gene Aliases
CELE_C42D8.5
Chromosome Location
Chr.X: 5092456 - 5097067 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029282.6 NP_001024453.1 10 - 11 2544 87

View Full Product Details