Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
vps-41
Entrez Gene ID
180800
Gene Name
protein-coding
Gene Aliases
CELE_F32A6.3
Chromosome Location
Chr.X: 5283831 - 5289731 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001380962.1 NP_001367568.1 6 - 7 1083 95
NM_001383575.1 6 - 7 1088 95
NM_001383576.1 4 - 5 657 95

View Full Product Details