Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sto-2
Entrez Gene ID
180802
Gene Name
protein-coding
Gene Aliases
CELE_F32A6.5
Chromosome Location
Chr.X: 5307486 - 5323387 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001270091.4 NP_001257020.1 8 - 9 894 68
NM_001270092.5 NP_001257021.1 8 - 9 2605 68
NM_001270093.4 NP_001257022.1 8 - 9 887 68
NM_001270094.2 NP_001257023.1 7 - 8 778 68

View Full Product Details