Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srv-7
Entrez Gene ID
259994
Gene Name
Serpentine Receptor, class V
Gene Aliases
CELE_T22B7.5
Chromosome Location
Chr.X: 5642856 - 5647984 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_076549.6 NP_508950.2 2 - 3 299 134

View Full Product Details