Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nas-33
Entrez Gene ID
186987
Gene Name
Zinc metalloproteinase nas-33
Gene Aliases
CELE_K04E7.3
Chromosome Location
Chr.X: 6451698 - 6455257 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_076685.4 NP_509086.2 8 - 9 882 67

View Full Product Details