Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ceh-60
Entrez Gene ID
184803
Gene Name
Homeobox domain-containing protein
Gene Aliases
CELE_F22A3.5
Chromosome Location
Chr.X: 6522596 - 6530148 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_076700.5 NP_509101.4 8 - 8 961 63

View Full Product Details