Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sur-5
Entrez Gene ID
180992
Gene Name
Acetoacetyl-CoA synthetase
Gene Aliases
CELE_K03A1.5
Chromosome Location
Chr.X: 7301329 - 7305864 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_076828.8 NP_509229.1 6 - 7 995 78
GenBank mRNA AY091467.1 - 5 - 6 926 78

View Full Product Details