Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
clc-2
Entrez Gene ID
180999
Gene Name
protein-coding
Gene Aliases
CELE_C01C10.1
Chromosome Location
Chr.X: 7431465 - 7435732 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001375068.3 NP_001362078.1 6 - 6 505 79
NM_076856.6 NP_509257.1 6 - 7 639 79
GenBank mRNA AF252604.1 - 6 - 7 641 79

View Full Product Details