Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- unc-98
- Entrez Gene ID
-
181020
- Gene Name
- Zinc finger protein unc-98
- Gene Aliases
-
CELE_F08C6.7
- Chromosome Location
-
Chr.X: 7570374 - 7573751 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_076883.7 |
NP_509284.2 |
6 - 7 |
783 |
70 |
GenBank mRNA |
AF515600.1 |
- |
6 - 7 |
708 |
70 |
- Gene Symbol
- apy-1
- Entrez Gene ID
-
181019
- Gene Name
- Apyrase apy-1
- Gene Aliases
-
CELE_F08C6.6
- Chromosome Location
-
Chr.X: 7568125 - 7573625 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_076882.10 |
NP_509283.1 |
6 - 7 |
657 |
70 |
View Full Product Details