Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
stn-2
Entrez Gene ID
181029
Gene Name
PDZ domain-containing protein
Gene Aliases
CELE_F27D9.8
Chromosome Location
Chr.X: 7676511 - 7680636 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029439.4 NP_001024610.1 11 - 12 1158 111

View Full Product Details