Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ref-2
Entrez Gene ID
183539
Gene Name
protein-coding
Gene Aliases
CELE_C47C12.3
Chromosome Location
Chr.X: 7744239 - 7750013 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029306.4 NP_001024477.1 4 - 5 493 99
NM_001029307.5 NP_001024478.1 5 - 6 524 99

View Full Product Details